4-Fluoro-2-iodotoluene ≥97.0%

Supplier: TCI America

Synonyms: 1-Fluoro-3-iodo-4-methylbenzene, 4-Fluoro-2-iodo-1-methylbenzene

F0613-25G F0613-5G
TCF0613-25GEA 154 USD
TCF0613-25G TCF0613-5G
4-Fluoro-2-iodotoluene ≥97.0%
4-Fluoro-2-iodotoluene

CAS Number: 13194-67-7
MDL Number: MFCD00013709
Molecular Formula: C7H6FI
Molecular Weight: 236.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 87
Specific Gravity (20/20): 1.81

Formula: C₇H₆FI
Boiling Pt: 90 °C
Density: 1.81 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00013709
CAS Number: 13194-67-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bb58837890336520a2f46810fd04ed278d857e&vid=70f4507387225099c93bba3432945abdf39c7fac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR