2-Fluoro-5-methylpyridine ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 2369-19-9
MDL Number: MFCD00041225
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 48
Specific Gravity (20/20): 1.09
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoro-3-picoline
Formula:
C₆H₆FN Boiling Pt: 155 °C Density: 1.09 (20 °C) Flash Pt: 48 °C |
MDL Number:
MFCD00041225 CAS Number: 2369-19-9 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...