2-Fluoro-4-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-Fluoro-4-picoline

F0579-25G F0579-5G
TCF0579-25GEA 81.6 USD
TCF0579-25G TCF0579-5G
2-Fluoro-4-methylpyridine ≥98.0%
2-Fluoro-4-methylpyridine

CAS Number: 461-87-0
MDL Number: MFCD00041224
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 51
Specific Gravity (20/20): 1.09


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-4-picoline

Formula: C₆H₆FN
Boiling Pt: 157 °C
Density: 1.09 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00041224
CAS Number: 461-87-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5715b91e534328a7bf5fbf9a3eb00a8eb26e002f&vid=5715b91e534328a7bf5fbf9a3eb00a8eb26e002f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR