2-Fluoro-5-nitrobenzonitrile ≥97.0% (by GC)

Supplier: TCI America

F0550-25G F0550-5G
TCF0550-25GEA 174.7 USD
TCF0550-25G TCF0550-5G
2-Fluoro-5-nitrobenzonitrile ≥97.0% (by GC)
2-Fluoro-5-nitrobenzonitrile

CAS Number: 17417-09-3
MDL Number: MFCD00042299
Molecular Formula: C7H3FN2O2
Molecular Weight: 166.11
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 94
Melting point (°C): 73

Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Boiling Pt: 94 °C
Melting Pt: 73 °C
MDL Number: MFCD00042299
CAS Number: 17417-09-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cb64579b5c42608156a2b496d523f3873b0d688&vid=6cb64579b5c42608156a2b496d523f3873b0d688&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR