4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
Supplier: TCI America
F0544-1G
F0544-5G
TCF0544-1GEA
34
USD
TCF0544-1G
TCF0544-5G
4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
4-Fluoro-2-(trifluoromethyl)benzaldehyde
CAS Number: 90176-80-0
Molecular Formula: C8H4F4O
Molecular Weight: 192.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 57
Specific Gravity (20/20): 1.42
Formula:
C₈H₄F₄O Boiling Pt: 114 °C Density: 1.42 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00061313 CAS Number: 90176-80-0 UN: 1989 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...