3-Fluorobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (3-Fluorophenyl)methanamine

F0539-25G F0539-5G
TCF0539-25GEA 196 USD
TCF0539-25G TCF0539-5G
3-Fluorobenzylamine ≥98.0% (by GC, titration analysis)
3-Fluorobenzylamine

CAS Number: 100-82-3
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 71
Specific Gravity (20/20): 1.11

Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 82 °C
Density: 1.11 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00008113
CAS Number: 100-82-3
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73ddc8259dfc465e1679119a306a223f86b5a6c0&vid=73ddc8259dfc465e1679119a306a223f86b5a6c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR