2-Fluoro-3-methylpyridine ≥98.0% (by GC)

Supplier: TCI America

F0533-1G F0533-5G
TCF0533-1GEA 30 USD
TCF0533-1G TCF0533-5G
2-Fluoro-3-methylpyridine ≥98.0% (by GC)
2-Fluoro-3-methylpyridine

CAS Number: 2369-18-8
MDL Number: MFCD00041223
Molecular Formula: C6H6FN
Molecular Weight: 111.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 48
Specific Gravity (20/20): 1.10


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-3-picoline

Formula: C₆H₆FN
Boiling Pt: 151 °C
Density: 1.10 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00041223
CAS Number: 2369-18-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6064a554c451ad7409e92ac21303bff02ca60ba&vid=e6064a554c451ad7409e92ac21303bff02ca60ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR