2-Fluoro-p-toluidine ≥97.0%
Supplier: TCI America
Synonyms:
2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene
F0529-25G
F0529-5G
TCF0529-25GEA
295
USD
TCF0529-25G
TCF0529-5G
2-Fluoro-p-toluidine ≥97.0%
2-Fluoro-p-toluidine
CAS Number: 452-80-2
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 71
Melting point (°C): 3
Flash Point (°C): 79
Specific Gravity (20/20): 1.11
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Amino-3-fluorotoluene, 2-Fluoro-p-toluidine
Formula:
C₇H₈FN Boiling Pt: 71 °C Melting Pt: 3 °C Density: 1.11 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00040975 CAS Number: 452-80-2 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...