4-Fluorocatechol ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 367-32-8
MDL Number: MFCD01317539
Molecular Formula: C6H5FO2
Molecular Weight: 128.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol
Formula:
C₆H₅FO₂ Melting Pt: 91 °C |
MDL Number:
MFCD01317539 CAS Number: 367-32-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...