Methyl-3-furoate ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Methyl furan-3-carboxylate

F0468-25G F0468-5G
TCF0468-25GEA 375 USD
TCF0468-25G TCF0468-5G
Methyl-3-furoate ≥96.0% (by GC)
Methyl-3-furoate

CAS Number: 13129-23-2
MDL Number: MFCD06203671
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 3-Furoic Acid Methyl Ester, 3-Furancarboxylic Acid Methyl Ester, Methyl 3-Furoate

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 64 °C
Density: 1.17 (20 °C)
MDL Number: MFCD06203671
CAS Number: 13129-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f923cfd74ef0ebd1afffb7a23ff772c7877cc15&vid=5f923cfd74ef0ebd1afffb7a23ff772c7877cc15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR