Ethyl-3-furoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl furan-3-carboxylate, Ethyl 3-Furoate, Ethyl 3-Furancarboxylate, 3-Furancarboxylic acid ethyl ester, 3-Furoic acid ethyl ester

F0463-25G F0463-5G
TCF0463-25GEA 163 USD
TCF0463-25G TCF0463-5G
Ethyl-3-furoate ≥98.0% (by GC)
Ethyl-3-furoate

CAS Number: 614-98-2
MDL Number: MFCD00005348
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 59
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Ethyl 3-Furoate, 3-Furancarboxylic Acid Ethyl Ester, 3-Furoic Acid Ethyl Ester

Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 95 °C
Density: 1.11 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00005348
CAS Number: 614-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28ea0beb05ab9012fd699101c694a9d1dac42eee&vid=28ea0beb05ab9012fd699101c694a9d1dac42eee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR