3-Fluorostyrene ≥97.0% (by GC) stabilized

Supplier: TCI America

F0409-1G F0409-5G
TCF0409-1GEA 62 USD
TCF0409-1G TCF0409-5G
3-Fluorostyrene ≥97.0% (by GC) stabilized
3-Fluorostyrene

(stabilized with TBC)
CAS Number: 350-51-6
MDL Number: MFCD00000341
Molecular Formula: C8H7F
Molecular Weight: 122.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 41
Flash Point (°C): 29
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C

Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 41 °C
Density: 1.03 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00000341
CAS Number: 350-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d16cdc45b7ce100c2960a6b4b587e9b00251b1d&vid=7d16cdc45b7ce100c2960a6b4b587e9b00251b1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR