2-Fluoro-m-xylene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Fluoro-1,3-dimethylbenzene, 1-Fluoro-2,6-dimethylbenzene
F0402-25G
F0402-5G
TCF0402-25GEA
190
USD
TCF0402-25G
TCF0402-5G
2-Fluoro-m-xylene ≥98.0% (by GC)
2-Fluoro-m-xylene
CAS Number: 443-88-9
MDL Number: MFCD00039215
Molecular Formula: C8H9F
Molecular Weight: 124.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 31
Specific Gravity (20/20): 0.99
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,6-Dimethylfluorobenzene, 2-Fluoro-1,3-dimethylbenzene
Formula:
C₈H₉F MW: 124.16 g/mol Boiling Pt: 167 °C Density: 0.99 (20 °C) Flash Pt: 31 °C |
MDL Number:
MFCD00039215 CAS Number: 443-88-9 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...