α,α,α,3-Tetrafluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Fluorobenzotrifluoride, 1-Fluoro-3-(trifluoromethyl)benzene

F0390-250G F0390-25G
TCF0390-250GEA 402 USD
TCF0390-250G TCF0390-25G
α,α,α,3-Tetrafluorotoluene ≥98.0% (by GC)
α,α,α,3-Tetrafluorotoluene

CAS Number: 401-80-9
MDL Number: MFCD00000382
Molecular Formula: C7H4F4
Molecular Weight: 164.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 7
Specific Gravity (20/20): 1.30


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Fluoro-3-(trifluoromethyl)benzene, alpha,alpha,alpha,3-Tetrafluorotoluene

Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 101 °C
Density: 1.30 (20 °C)
Flash Pt: 7 °C
MDL Number: MFCD00000382
CAS Number: 401-80-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8baa9c51e01ff2704905032f8d33df0bcd48feaa&vid=cc4a5cc86ff959f1973fe62e78a7e4ea2de498c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR