2-Fluoro-α-methyl-4-biphenylacetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
CAS Number: 5104-49-4
MDL Number: MFCD00079303
Molecular Formula: C15H13FO2
Molecular Weight: 244.27
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 113
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Fluorinated Building Blocks, Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredie
Other Product Names / Synonyms: 2-(2-Fluorobiphenyl-4-yl)propionic Acid
Formula:
C₁₅H₁₃FO₂ MW: 244.265 g/mol Melting Pt: 113 °C |
MDL Number:
MFCD00079303 CAS Number: 5104-49-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (T,HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...