2'-Fluoroacetophenone ≥97.0% (by GC)

Supplier: TCI America

F0364-25G F0364-5G
TCF0364-25GEA 26 USD
TCF0364-25G TCF0364-5G
2'-Fluoroacetophenone ≥97.0% (by GC)
2'-Fluoroacetophenone

CAS Number: 445-27-2
MDL Number: MFCD00000320
Molecular Formula: C8H7FO
Molecular Weight: 138.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 61
Specific Gravity (20/20): 1.14

Formula: C₈H₇FO
Boiling Pt: 85 °C
Density: 1.14 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00000320
CAS Number: 445-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c7312bd377c6fef0107ddc0c3841623a03a2413&vid=2c7312bd377c6fef0107ddc0c3841623a03a2413&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR