4-Fluoro-3-nitrobenzotrifluoride ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 4-Fluoro-3-Nitrobenzotrifluoride

F0324-250G F0324-25G
TCF0324-250GEA 107 USD
TCF0324-250G TCF0324-25G
4-Fluoro-3-nitrobenzotrifluoride ≥95.0% (by GC)
4-Fluoro-3-nitrobenzotrifluoride

CAS Number: 367-86-2
MDL Number: MFCD00007059
Molecular Formula: C7H3F4NO2
Molecular Weight: 209.10
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 33
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Fluoro-2-nitro-4-(trifluoromethyl)benzene, 3-Nitro-alpha,alpha,alpha,4-tetrafluorotoluene

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Boiling Pt: 208 °C
Density: 1.53 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00007059
CAS Number: 367-86-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96b02c80ffd46186c5320b57289d415c25eeb10&vid=d38a8b7343a38635d9a3e5a6fbdca5234195b453&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR