Diethyl fluoromalonate ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
1,3-diethyl 2-fluoropropanedioate
F0283-1G
F0283-25G
F0283-5G
TCF0283-1GEA
34
USD
TCF0283-1G
TCF0283-25G
TCF0283-5G
Diethyl fluoromalonate ≥95.0% (by GC)
Diethyl fluoromalonate
CAS Number: 685-88-1
MDL Number: MFCD00009139
Molecular Formula: C7H11FO4
Molecular Weight: 178.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 62
Specific Gravity (20/20): 1.15
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Fluoromalonic Acid Diethyl Ester
Formula:
C₇H₁₁FO₄ MW: 178.16 g/mol Boiling Pt: 122 °C Density: 1.15 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00009139 CAS Number: 685-88-1 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...