3-Fluoropyrocatechol ≥98.0% (by GC), Technical Grade
Supplier: TCI America
Synonyms:
3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
F0282-1G
F0282-5G
TCF0282-1GEA
47
USD
TCF0282-1G
TCF0282-5G
3-Fluoropyrocatechol ≥98.0% (by GC), Technical Grade
3-Fluoropyrocatechol
CAS Number: 363-52-0
MDL Number: MFCD00042582
Molecular Formula: C6H5FO2
Molecular Weight: 128.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Fluoro-2,3-dihydroxybenzene
Formula:
C₆H₅FO₂ MW: 128.1 g/mol Melting Pt: 72 °C |
MDL Number:
MFCD00042582 CAS Number: 363-52-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...