3-Fluorobenzoyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

F0277-25G F0277-5G
TCF0277-25GEA 87 USD
TCF0277-25G TCF0277-5G
3-Fluorobenzoyl chloride ≥97.0% (by GC, titration analysis)
3-Fluorobenzoyl chloride

CAS Number: 1711-07-5
MDL Number: MFCD00000670
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 91
Melting point (°C): -30
Flash Point (°C): 82
Specific Gravity (20/20): 1.32

Formula: C₇H₄ClFO
Boiling Pt: 91 °C
Melting Pt: –30 °C
Density: 1.32 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00000670
CAS Number: 1711-07-5
EINECS: 216-977-3
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f04a37933fd9b34ed57d1457ec8f3c523b0d1a98&vid=cae9e880f5d05e1f14e29a4a24bbdcfc46a72799&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR