3'-Fluoroacetophenone ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-(3-Fluorophenyl)ethanone, m-Fluoroacetophenone

F0262-25G F0262-5G
TCF0262-25GEA 169 USD
TCF0262-25G TCF0262-5G
3'-Fluoroacetophenone ≥97.0% (by GC)
3'-Fluoroacetophenone

CAS Number: 455-36-7
MDL Number: MFCD00000338
Molecular Formula: C8H7FO
Molecular Weight: 138.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Melting point (°C): -3
Flash Point (°C): 80
Specific Gravity (20/20): 1.14

Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 81 °C
Melting Pt: –3 °C
Density: 1.14 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00000338
CAS Number: 455-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0dda57f76fc4a9b2a926fc15f035fb70547850&vid=ea0dda57f76fc4a9b2a926fc15f035fb70547850&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR