(4-Fluorophenyl)acetonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 459-22-3
MDL Number: MFCD00001917
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 108
Specific Gravity (20/20): 1.14
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluorophenylacetonitrile
Formula:
C₈H₆FN Boiling Pt: 230 °C Density: 1.14 (20 °C) Flash Pt: 108 °C |
MDL Number:
MFCD00001917 CAS Number: 459-22-3 UN: 3276 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...