4-Fluoro-ɑ-methylstyrene ≥98.0% (by GC) stabilized

Supplier: TCI America

Synonyms: 1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene

F0250-25ML F0250-5ML
TCF0250-25MLEA 250 USD
TCF0250-25ML TCF0250-5ML
4-Fluoro-ɑ-methylstyrene ≥98.0% (by GC) stabilized
4-Fluoro-alpha-methylstyrene

(stabilized with TBC)
CAS Number: 350-40-3
MDL Number: MFCD00042297
Molecular Formula: C9H9F
Molecular Weight: 136.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 56
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-(4-Fluorophenyl)-1-propene

Formula: C₉H₉F
MW: 136.17 g/mol
Boiling Pt: 183 °C
Density: 1.02 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00042297
CAS Number: 350-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95daaa6aa7fb9fa462ba3f9982fdcf09cfae5515&vid=95daaa6aa7fb9fa462ba3f9982fdcf09cfae5515&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR