3-Fluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
F0219-25G
F0219-5G
TCF0219-25GEA
132.9
USD
TCF0219-25G
TCF0219-5G
3-Fluorobenzonitrile ≥98.0% (by GC)
3-Fluorobenzonitrile
CAS Number: 403-54-3
MDL Number: MFCD00001797
Molecular Formula: C7H4FN
Molecular Weight: 121.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 68
Freezing point (°C): 15
Specific Gravity (20/20): 1.14
Formula:
C₇H₄FN MW: 121.11 g/mol Boiling Pt: 183 °C Density: 1.14 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00001797 CAS Number: 403-54-3 UN: 3276 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...