α-Bromo-4-fluorotoulene ≥97.0%

Supplier: TCI America

Synonyms: α-Bromo-p-fluorotoulene, 4-Fluorobenzyl bromide, p-Fluorobenzyl bromide

F0214-25G F0214-5G
TCF0214-25GEA 87.53 USD
TCF0214-25G TCF0214-5G
α-Bromo-4-fluorotoulene ≥97.0%
α-Bromo-4-fluorotoulene

CAS Number: 459-46-1
MDL Number: MFCD00000359
Molecular Formula: C7H6BrF
Molecular Weight: 189.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 113
Specific Gravity (20/20): 1.54

Formula: C₇H₆BrF
Boiling Pt: 177 °C
Density: 1.54 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000359
CAS Number: 459-46-1
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ef01174c8e977c7aabf3cfd8726da8e18bd9398&vid=8ee4aab11daff7c0845a6a7212a844b05f893e01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR