α-Chloro-3-fluorotoulene ≥95.0% (by GC)
Supplier: TCI America
F0210-25G
F0210-5G
TCF0210-25GEA
62.8
USD
TCF0210-25G
TCF0210-5G
α-Chloro-3-fluorotoulene ≥95.0% (by GC)
α-Chloro-3-fluorotoulene
CAS Number: 456-42-8
MDL Number: MFCD00000904
Molecular Formula: C7H6ClF
Molecular Weight: 144.57
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 58
Specific Gravity (20/20): 1.22
Formula:
C₇H₆ClF Boiling Pt: 180 °C Density: 1.22 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00000904 CAS Number: 456-42-8 UN: 2920 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...