3-Fluoropyridine ≥98.0% (by GC)
Supplier: TCI America
F0207-25G
F0207-5G
TCF0207-25GEA
184
USD
TCF0207-25G
TCF0207-5G
3-Fluoropyridine ≥98.0% (by GC)
3-Fluoropyridine
CAS Number: 372-47-4
MDL Number: MFCD00006374
Molecular Formula: C5H4FN
Molecular Weight: 97.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 13
Specific Gravity (20/20): 1.14
Formula:
C₅H₄FN MW: 97.09 g/mol Boiling Pt: 106 °C Density: 1.14 (20 °C) Flash Pt: 13 °C |
MDL Number:
MFCD00006374 CAS Number: 372-47-4 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...