4-Fluorobenzenesulfonyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Fluorobenzene-1-sulphonyl chloride

F0202-25G F0202-5G
TCF0202-25GEA 57.98 USD
TCF0202-25G TCF0202-5G
4-Fluorobenzenesulfonyl chloride ≥97.0% (by GC, titration analysis)
4-Fluorobenzenesulphonyl chloride

CAS Number: 349-88-2
MDL Number: MFCD00007438
Molecular Formula: C6H4ClFO2S
Molecular Weight: 194.60
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 35
Flash Point (°C): 110

Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
Boiling Pt: 112 °C
Melting Pt: 35 °C
Flash Pt: 110 °C
MDL Number: MFCD00007438
CAS Number: 349-88-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47afa8821206110b5bfc5c21f6fe89ba4e3d98b3&vid=47afa8821206110b5bfc5c21f6fe89ba4e3d98b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR