9-Fluorenylmethyl chloroformate (Fmoc chloride) ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

F0197-100G F0197-25G F0197-5G
TCF0197-100GEA 499 USD
TCF0197-100G TCF0197-25G TCF0197-5G
9-Fluorenylmethyl chloroformate (Fmoc chloride) ≥97.0% (by HPLC, titration analysis)
9-Fluorenylmethyl chloroformate (Fmoc chloride)

[N-Protecting Agent for Peptides Research]
CAS Number: 28920-43-6
MDL Number: MFCD00001138
Molecular Formula: C15H11ClO2
Molecular Weight: 258.70
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 62
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents, Analytical Science, Fluorescence Detection (HPLC Labeling Reagents), UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Fmoc-Cl, Chloroformic Acid 9-Fluorenylmethyl Ester, (9H-Fluoren-9-ylmethoxy)carbonyl Chloride

Formula: C₁₅H₁₁ClO₂
MW: 258.7 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00001138
CAS Number: 28920-43-6
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c750a79625ecbc6892647a3bf340bda1fa649a5&vid=ab73037376df8c6d2360f1c11db5772121b13509&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR