Fumaroyl chloride ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Fumaryl chloride

F0152-25G F0152-500G
TCF0152-25GEA 49.2 USD
TCF0152-25G TCF0152-500G
Fumaroyl chloride ≥95.0% (by GC, titration analysis)
Fumaroyl chloride

CAS Number: 627-63-4
MDL Number: MFCD00000733
Molecular Formula: C4H2Cl2O2
Molecular Weight: 152.96
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 164
Melting point (°C): -2
Flash Point (°C): 77
Specific Gravity (20/20): 1.41

Formula: C₄H₂Cl₂O₂
MW: 152.96 g/mol
Boiling Pt: 164 °C
Melting Pt: –2 °C
Density: 1.41 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00000733
CAS Number: 627-63-4
UN: 1780
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5052ce0a769231068e6255ba8deded48fc9bb3&vid=da5052ce0a769231068e6255ba8deded48fc9bb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR