5-Fluorouracil ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
CAS Number: 51-21-8
MDL Number: MFCD00006018
Molecular Formula: C4H3FN2O2
Molecular Weight: 130.08
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 283
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Reagents for Central Nervous System Disease Models, Reagents for Inflammation & Immunity Disorder Models, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases
Other Product Names / Synonyms: 5-FU
Formula:
C₄H₃FN₂O₂ MW: 130.08 g/mol Melting Pt: 283 °C |
MDL Number:
MFCD00006018 CAS Number: 51-21-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...