Fumaronitrile ≥98.0% (by GC)
Supplier: TCI America
F0072-25G
F0072-5G
TCF0072-25GEA
185
USD
TCF0072-25G
TCF0072-5G
Fumaronitrile ≥98.0% (by GC)
Fumaronitrile
CAS Number: 764-42-1
MDL Number: MFCD00001928
Molecular Formula: C4H2N2
Molecular Weight: 78.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 186
Melting point (°C): 97
Formula:
C₄H₂N₂ MW: 78.07 g/mol Boiling Pt: 186 °C Melting Pt: 97 °C |
MDL Number:
MFCD00001928 CAS Number: 764-42-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...