Ethyl isobutyrylacetate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl 4-methyl-3-oxopentanoate

E0882-25G E0882-5G
TCE0882-25GEA 152 USD
TCE0882-25G TCE0882-5G
Ethyl isobutyrylacetate ≥95.0%
Ethyl isobutyrylacetate

CAS Number: 7152-15-0
MDL Number: MFCD00009198
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 44
Specific Gravity (20/20): 0.98


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Isobutyrylacetic Acid Ethyl Ester, Ethyl 4-Methyl-3-oxovalerate, 4-Methyl-3-oxovaleric Acid Ethyl Ester

Formula: C₈H₁₄O₃
Boiling Pt: 90 °C
Density: 0.98 (20 °C)
Flash Pt: 44 °C
MDL Number: MFCD00009198
CAS Number: 7152-15-0
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21566831159f30757304db6c2f359617e2f98b1&vid=b21566831159f30757304db6c2f359617e2f98b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR