Ethyl-3-cyclopentene-1-carboxylate ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Ethyl cyclopent-3-enecarboxylate, 3-Cyclopentene-1-carboxylic Acid Ethyl Ester
E0831-25G
E0831-5G
TCE0831-25GEA
268
USD
TCE0831-25G
TCE0831-5G
Ethyl-3-cyclopentene-1-carboxylate ≥97.0% (by GC)
Ethyl-3-cyclopentene-1-carboxylate
CAS Number: 21622-01-5
MDL Number: MFCD06410819
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 3-Cyclopentene-1-carboxylic Acid Ethyl Ester
Formula:
C₈H₁₂O₂ MW: 140.18 g/mol Boiling Pt: 64 °C Density: 1.00 (20 °C) |
MDL Number:
MFCD06410819 CAS Number: 21622-01-5 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...