Ethyl-4,4,4-trifluorocrotonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl (E)-4,4,4-trifluorobut-2-enoate, Ethyl 4,4,4-trifluorocrotonate

E0772-25G E0772-5G
TCE0772-25GEA 281.4 USD
TCE0772-25G TCE0772-5G
Ethyl-4,4,4-trifluorocrotonate ≥98.0% (by GC)
Ethyl 4,4,4-trifluorocrotonate

CAS Number: 25597-16-4
MDL Number: MFCD00009903
Molecular Formula: C6H7F3O2
Molecular Weight: 168.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 24
Specific Gravity (20/20): 1.17


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 4,4,4-Trifluorocrotonic Acid Ethyl Ester

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 115 °C
Density: 1.17 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00009903
CAS Number: 25597-16-4
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d45b4df36069f1f0b6431bbb490289f04921ee1b&vid=d45b4df36069f1f0b6431bbb490289f04921ee1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR