4-Ethylphenyl isocyanate ≥97.0% (by GC)

Supplier: TCI America

E0767-1G E0767-5G
TCE0767-1GEA 27 USD
TCE0767-1G TCE0767-5G
4-Ethylphenyl isocyanate ≥97.0% (by GC)
4-Ethylphenyl isocyanate

CAS Number: 23138-50-3
MDL Number: MFCD00013881
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 88
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Isocyanic Acid 4-Ethylphenyl Ester

Formula: C₉H₉NO
Boiling Pt: 80 °C
Density: 1.04 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00013881
CAS Number: 23138-50-3
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d602ff099111fc9babce76a4a3c9c42698633611&vid=d602ff099111fc9babce76a4a3c9c42698633611&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR