1-Ethyl-3-methylimidazolium methanesulfonate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

E0755-25G E0755-5G
TCE0755-25GEA 83 USD
TCE0755-25G TCE0755-5G
1-Ethyl-3-methylimidazolium methanesulfonate ≥98.0% (by total nitrogen basis)
1-Ethyl-3-methylimidazolium methanesulphonate

CAS Number: 145022-45-3
MDL Number: MFCD06798171
Molecular Formula: C7H14N2O3S
Molecular Weight: 206.26
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Flash Point (°C): 67
Specific Gravity (20/20): 1.25

Formula: C₇H₁₄N₂O₃S
Density: 1.25 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD06798171
CAS Number: 145022-45-3
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2eeab0ac1f3c777352c7eed75518e36fc0326b0&vid=a2eeab0ac1f3c777352c7eed75518e36fc0326b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR