7-Ethyl-10-hydroxycamptothecin ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 86639-52-3
MDL Number: MFCD06762720
Molecular Formula: C22H20N2O5
Molecular Weight: 392.41
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 217
Specific rotation [a]20/D: 21.5 deg (C=0.2, THF)
Product Categories: Bioscience, Antitumor Ingredients, Cancer Research Reagents, Quinoline Alkaloids
Other Product Names / Synonyms: SN-38
Formula:
C22H20N2O5 Melting Pt: 217 °C |
MDL Number:
MFCD00871873 CAS Number: 86639-52-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...