3,4-Ethylenedioxythiophene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,3-Dihydrothieno[3,4-b][1,4]dioxine, EDOT
CAS Number: 126213-50-1
MDL Number: MFCD02093622
Molecular Formula: C6H6O2S
Molecular Weight: 142.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 104
Freezing point (°C): 10
Specific Gravity (20/20): 1.34
Lambda max.: 260 nm (CH2Cl2)Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2,3-Dihydrothieno[3,4-b][1,4]dioxine, EDOT
Formula:
C₆H₆O₂S MW: 142.17 g/mol Boiling Pt: 112 °C Density: 1.34 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD02093622 CAS Number: 126213-50-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...