α-Chloro-4-ethyltoulene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Chloromethyl)-4-ethylbenzene, α-Chloro-p-ethyltoulene, 4-Ethylbenzyl chloride

E0738-25G E0738-5G
TCE0738-25GEA 223 USD
TCE0738-25G TCE0738-5G
α-Chloro-4-ethyltoulene ≥98.0% (by GC)
α-Chloro-4-ethyltoulene

CAS Number: 1467-05-6
MDL Number: MFCD00039357
Molecular Formula: C9H11Cl
Molecular Weight: 154.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 1.04

Formula: C₉H₁₁Cl
MW: 154.64 g/mol
Boiling Pt: 111 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00039357
CAS Number: 1467-05-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14d9b609e842ee358fcf9c04064fbb6c446db071&vid=14d9b609e842ee358fcf9c04064fbb6c446db071&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR