Ethyl-4-cyanocinnamate ≥98.0% (by GC)

Supplier: TCI America

E0677-1G E0677-5G
TCE0677-1GEA 106 USD
TCE0677-1G TCE0677-5G
Ethyl-4-cyanocinnamate ≥98.0% (by GC)
Ethyl-4-cyanocinnamate

CAS Number: 62174-99-6
Molecular Formula: C12H11NO2
Molecular Weight: 201.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 4-Cyanocinnamic Acid Ethyl Ester

Formula: C₁₂H₁₁NO₂
Melting Pt: 70 °C
CAS Number: 62174-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc6494fe5de20775ead619634b192ea21adaa3b4&vid=bc6494fe5de20775ead619634b192ea21adaa3b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR