3-Ethynyltoluene ≥98.0%

Supplier: TCI America

Synonyms: 1-Ethynyl-3-methylbenzene, m-Tolylacetylene

E0665-1G E0665-25G E0665-5G
TCE0665-1GEA 39 USD
TCE0665-1G TCE0665-25G TCE0665-5G
3-Ethynyltoluene ≥98.0%
3-Ethynyltoluene

CAS Number: 766-82-5
Molecular Formula: C9H8
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 25
Specific Gravity (20/20): 0.92


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 1-Ethynyl-3-methylbenzene, m-Tolylacetylene

Formula: C₉H₈
MW: 116.16 g/mol
Boiling Pt: 60 °C
Density: 0.92 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD01318174
CAS Number: 766-82-5
UN: 3295
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85f4ef73a3da6bd8b7078711838ca3792b3e5d32&vid=85f4ef73a3da6bd8b7078711838ca3792b3e5d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR