Ethyl-2-fluoropropionate ≥97.0%
Supplier: TCI America
Synonyms:
2-Fluoropropionic Acid Ethyl Ester, Ethyl 2-fluoropropionate
E0663-25G
E0663-5G
TCE0663-25GEA
128
USD
TCE0663-25G
TCE0663-5G
Ethyl-2-fluoropropionate ≥97.0%
Ethyl-2-fluoropropionate
CAS Number: 349-43-9
MDL Number: MFCD01320801
Molecular Formula: C5H9FO2
Molecular Weight: 120.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Specific Gravity (20/20): 1.03
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoropropionic Acid Ethyl Ester
Formula:
C₅H₉FO₂ Boiling Pt: 121 °C Density: 1.03 (20 °C) |
MDL Number:
MFCD01320801 CAS Number: 349-43-9 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...