4-Ethynyltoluene 98,0
Supplier: TCI America
Synonyms:
1-Ethynyl-4-methylbenzene, p-Tolylacetylene
E0655-25G
E0655-5G
TCE0655-25GEA
176
USD
TCE0655-25G
TCE0655-5G
4-Ethynyltoluene 98,0
4-Ethynyltoluene
CAS Number: 766-97-2
Molecular Formula: C9H8
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 168
Flash Point (°C): 48
Freezing point (°C): 23
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 1-Ethynyl-4-methylbenzene, p-Tolylacetylene
Formula:
C₉H₈ MW: 116.16 g/mol Boiling Pt: 168 °C Flash Pt: 48 °C |
MDL Number:
MFCD00008571 CAS Number: 766-97-2 UN: 3295 ADR: 3 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...