(R)-(-)-Epichlorhydrin ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane

E0581-25G E0581-5G
TCE0581-25GEA 55.31 USD
TCE0581-25G TCE0581-5G
(R)-(-)-Epichlorhydrin ≥98.0% (by GC)
(R)-(-)-Epichlorhydrin

CAS Number: 51594-55-9
MDL Number: MFCD00077759
Molecular Formula: C3H5ClO
Molecular Weight: 92.52
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 31
Specific Gravity (20/20): 1.18
Specific rotation [a]20/D: -44.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Chloro-2,3-epoxypropane, (R)-Chloromethyloxirane, (R)-3-Chloropropylene Oxide

Formula: ClCH₂CH(O)CH₂
MW: 92.52 g/mol
Boiling Pt: 116 °C
Density: 1.18 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00077759
CAS Number: 51594-55-9
UN: 2023
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=074403a3e3aee2df9bd44a35f4bc27eae5010c9a&vid=aa6b693d37f853a6cdbb5bd70b1fdf9e3ffce757&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR