3-Ethynylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0560-1G E0560-5G
TCE0560-1GEA 45 USD
TCE0560-1G TCE0560-5G
3-Ethynylpyridine ≥98.0% (by GC, titration analysis)
3-Ethynylpyridine

CAS Number: 2510-23-8
MDL Number: MFCD02177459
Molecular Formula: C7H5N
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 36
Flash Point (°C): 57
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 3-Pyridylacetylene

Formula: C₇H₅N
Melting Pt: 36 °C
Flash Pt: 57 °C
MDL Number: MFCD02177459
CAS Number: 2510-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ef6cf5e0bc7ebb169acc701c8a9c89083693b2f&vid=5ef6cf5e0bc7ebb169acc701c8a9c89083693b2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR