(Ethyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
E0549-250G
E0549-25G
TCE0549-250GEA
318
USD
TCE0549-250G
TCE0549-25G
(Ethyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
(Ethyl)triphenylphosphonium iodide
CAS Number: 4736-60-1
MDL Number: MFCD00040352
Molecular Formula: C20H20IP
Molecular Weight: 418.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 170
Formula:
C₂₀H₂₀IP Melting Pt: 170 °C |
MDL Number:
MFCD00040352 CAS Number: 4736-60-1 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...