(S)-(+)-Epichlorhydrin ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(+)-Glycerol epichlorhydrin, (S)-(+)-3-Chloro-1,2-epoxypropane, (S)-(+)-1-Chloro-2,3-epoxypropane, (2S)-2-(chloromethyl)oxirane

E0533-25G E0533-5G
TCE0533-25GEA 67.39 USD
TCE0533-25G TCE0533-5G
(S)-(+)-Epichlorhydrin ≥98.0% (by GC)
(S)-(+)-Epichlorhydrin

CAS Number: 67843-74-7
MDL Number: MFCD00077760
Molecular Formula: C3H5ClO
Molecular Weight: 92.52
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 34
Specific Gravity (20/20): 1.18
Specific rotation [a]20/D: 45 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-Chloromethyloxirane, (S)-3-Chloropropylene Oxide, (S)-1-Chloro-2,3-epoxypropane

Formula: ClCH₂CH(O)CH₂
MW: 92.52 g/mol
Boiling Pt: 112 °C
Density: 1.18 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00077760
CAS Number: 67843-74-7
UN: 2023
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0e5049594662a0e0f9be4c990742dc65383fa81&vid=71aaf2806edadd3052725d5146e696e2ea1eafa5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR