2-Ethyl-4-methylthiazole ≥98.0% (by GC)

Supplier: TCI America

E0456-25G E0456-5G
TCE0456-25GEA 96 USD
TCE0456-25G TCE0456-5G
2-Ethyl-4-methylthiazole ≥98.0% (by GC)
2-Ethyl-4-methylthiazole

CAS Number: 15679-12-6 MDL Number: MFCD00036621 Molecular Formula: C6H9NS Molecular Weight: 127.21 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 67 Flash Point (°C): 54 Specific Gravity (20/20): 1.03

Formula: C₆H₉NS
MW: 127.21 g/mol
Boiling Pt: 67 °C
Density: 1.03 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00036621
CAS Number: 15679-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06ced2a30f6fdc7e0f7f32dd10196c20203ded79&vid=06ced2a30f6fdc7e0f7f32dd10196c20203ded79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR