5,6-Epoxy-1-hexene ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 1,2-Epoxy-5-hexene

E0410-25G E0410-5G
TCE0410-25GEA 206.5 USD
TCE0410-25G TCE0410-5G
5,6-Epoxy-1-hexene ≥96.0% (by GC)
5,6-Epoxy-1-hexene

CAS Number: 10353-53-4
MDL Number: MFCD00010051
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 15
Specific Gravity (20/20): 0.86

Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 121 °C
Density: 0.86 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00010051
CAS Number: 10353-53-4
UN: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fec2aa6ee98da0970d989d1cb2925ba93703f3f&vid=6fec2aa6ee98da0970d989d1cb2925ba93703f3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR